1,5-Dimethyl-1H-pyrazol-4-amine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1,5-Dimethyl-1H-pyrazol-4-amine xhydrochloride

A159143-1G A159143-5G
76812-622EA 58 USD
76812-622 76812-624
1,5-Dimethyl-1H-pyrazol-4-amine hydrochloride 95%
1,5-Dimethyl-1H-pyrazol-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClN₃
MW: 147.61 g/mol
MDL Number: MFCD11100176
CAS Number: 948571-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR