4-(((tert-Butoxycarbonyl)(methyl)amino)methyl)benzoic acid 97%

Supplier: Ambeed
Warning

A158949-1G A158949-250MG
76812-618EA 124.13 USD
76812-618 76812-620
4-(((tert-Butoxycarbonyl)(methyl)amino)methyl)benzoic acid 97%
4-(((tert-Butoxycarbonyl)(methyl)amino)methyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD17170958
CAS Number: 210963-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR