4-Bromo-3-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-Bromo-3-methylphenyl)boronic acid

A158721-100MG A158721-1G A158721-250MG A158721-25G A158721-5G
76812-608EA 13.8 USD
76812-608 76812-610 76812-612 76812-614 76812-616
4-Bromo-3-methylphenylboronic acid 97%
4-Bromo-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂
MW: 214.852 g/mol
MDL Number: MFCD17011983
CAS Number: 221006-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR