(2S)-2-Amino-2-phenylethanamide 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-2-phenylacetamide, (S)-(+)-2-Phenylglycine Amide

A158639-100G A158639-10G A158639-1G A158639-25G A158639-5G
76812-594EA 93 USD
76812-594 76812-596 76812-598 76812-600 76812-602
(2S)-2-Amino-2-phenylethanamide 97%
(2S)-2-Amino-2-phenylethanamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MDL Number: MFCD03791027
CAS Number: 6485-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR