1-Bromonaphthalen-2-amine 97%

Supplier: Ambeed
Warning

A158459-100MG A158459-1G A158459-250MG A158459-25G A158459-5G
76812-584EA 11 USD
76812-584 76812-586 76812-588 76812-590 76812-592
1-Bromonaphthalen-2-amine 97%
1-Bromonaphthalen-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MW: 222.08 g/mol
MDL Number: MFCD00086888
CAS Number: 20191-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR