(2-Phenylcyclopropyl)methanol 95%

Supplier: Ambeed
Warning

A158285-1G A158285-250MG A158285-100MG
76812-580EA 226 USD
76812-580 76812-582 77883-247
(2-Phenylcyclopropyl)methanol 95%
(2-Phenylcyclopropyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MDL Number: MFCD03931867
CAS Number: 61826-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR