2-Bromopyridine-3,4-diamine 97%

Supplier: Ambeed
Warning

A157943-100MG A157943-1G A157943-250MG A157943-5G
76812-556EA 37.14 USD
76812-556 76812-558 76812-560 77611-514
2-Bromopyridine-3,4-diamine 97%
2-Bromopyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MDL Number: MFCD12910485
CAS Number: 189230-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR