2,6-Difluoro-4-nitrophenol 97%

Supplier: Ambeed
Danger

A157052-100MG A157052-1G A157052-250MG A157052-5G A157052-10G A157052-25G
76812-498EA 13 USD
76812-498 76812-500 76812-502 77671-228 77883-237 77883-238
2,6-Difluoro-4-nitrophenol 97%
2,6-Difluoro-4-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₃
MDL Number: MFCD07366521
CAS Number: 658-07-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR