Methyl-2-(2-chloro-4-fluorophenyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-4-fluorophenylacetate, Methyl 2-(2-chloro-4-fluorophenyl)acetate

A157042-1G A157042-25G
76812-496EA 7 USD
76812-496 77671-224
Methyl-2-(2-chloro-4-fluorophenyl)acetate 98%
Methyl-2-(2-chloro-4-fluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MDL Number: MFCD00800611
CAS Number: 214262-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR