6-Bromo-1H-pyrrolo[3,2-b]pyridine-3-carbonitrile 97%

Supplier: Ambeed
Warning

A156629-100MG A156629-1G A156629-250MG
76812-478EA 37.14 USD
76812-478 76812-480 76812-482
6-Bromo-1H-pyrrolo[3,2-b]pyridine-3-carbonitrile 97%
6-Bromo-1H-pyrrolo[3,2-b]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrN₃
MW: 222.0400 g/mol
MDL Number: MFCD18380893
CAS Number: 1260386-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR