tert-Butyl piperidine-4-carboxylate 96%

Supplier: Ambeed
Warning

A156365-1G A156365-25G A156365-5G
76812-476EA 48.81 USD
76812-476 77549-984 77549-986
tert-Butyl piperidine-4-carboxylate 96%
tert-Butyl piperidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₂
MW: 185.263 g/mol
MDL Number: MFCD02684429
CAS Number: 138007-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR