N-(tert-Butyl)-1-methylcyclopropane-1-sulfonamide 97%

Supplier: Ambeed
Warning

A156340-1G A156340-250MG
76812-468EA 125.19 USD
76812-468 76812-470
N-(tert-Butyl)-1-methylcyclopropane-1-sulfonamide 97%
N-(tert-Butyl)-1-methylcyclopropane-1-sulfonamide

Precaución: for Research Use Only

Formula: C₈H₁₇NO₂S
MW: 191.29 g/mol
MDL Number: MFCD11707046
CAS Number: 669008-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR