(2-Amino-6-fluorophenyl)methanol 95%

Supplier: Ambeed
Warning

A156275-100MG A156275-1G A156275-250MG A156275-5G
76812-460EA 7 USD
76812-460 76812-462 76812-464 76812-466
(2-Amino-6-fluorophenyl)methanol 95%
(2-Amino-6-fluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.14 g/mol
MDL Number: MFCD12400884
CAS Number: 221285-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR