(2,4-Dichlorophenyl)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichlorobenzyl cyanide, 2-(2,4-Dichlorophenyl)acetonitrile

A155396-100G A155396-10G A155396-25G A155396-500G A155396-5G
76812-430EA 24 USD
76812-430 76812-432 76812-434 76812-436 76812-438
(2,4-Dichlorophenyl)acetonitrile 97%
(2,4-Dichlorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N
MDL Number: MFCD00001899
CAS Number: 6306-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR