3-Bromo-2-cyano-5-methylpyridine 97%

Supplier: Ambeed
Danger

A155336-250MG A155336-1G
76812-426EA 48 USD
76812-426 77883-221
3-Bromo-2-cyano-5-methylpyridine 97%
3-Bromo-2-cyano-5-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MDL Number: MFCD16988815
CAS Number: 474824-78-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR