3-(Trifluoromethyl)pyrazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A155317-100MG A155317-1G A155317-250MG
76812-420EA 33 USD
76812-420 76812-422 76812-424
3-(Trifluoromethyl)pyrazine-2-carboxylic acid 97%
3-(Trifluoromethyl)pyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃F₃N₂O₂
MDL Number: MFCD20646033
CAS Number: 870787-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR