5-Nitro-1H-indole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A155073-100G A155073-250MG A155073-25G A155073-5G A155073-10G A155073-1G
76812-410EA 531.01 USD
76812-410 76812-412 76812-414 76812-416 77883-217 77883-218
5-Nitro-1H-indole-3-carbaldehyde 98%
5-Nitro-1H-indole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C9H6N2O3
MDL Number: MFCD01313781
CAS Number: 6625-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR