7-Methoxyquinazolin-4(1H)-one 95%

Supplier: Ambeed
Warning

A155035-250MG A155035-5G A155035-100MG A155035-1G
76812-406EA 43.5 USD
76812-406 76812-408 77671-188 77671-190
7-Methoxyquinazolin-4(1H)-one 95%
7-Methoxyquinazolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.172 g/mol
MDL Number: MFCD08460971
CAS Number: 16064-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR