Ethyl ((benzyloxy)carbonyl)glycinate 97%

Supplier: Ambeed
Warning

A154211-100G A154211-10G A154211-1G A154211-250MG A154211-25G A154211-5G
76812-372EA 839.18 USD
76812-372 76812-374 76812-376 76812-378 76812-380 76812-382
Ethyl ((benzyloxy)carbonyl)glycinate 97%
Ethyl ((benzyloxy)carbonyl)glycinate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
MW: 237.25 g/mol
MDL Number: MFCD01724894
CAS Number: 1145-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR