6-Bromo-2-chloro-1,8-naphthyridine 98%

Supplier: Ambeed
Warning

A154029-250MG A154029-1G A154029-5G
76812-366EA 27.59 USD
76812-366 77611-458 77611-460
6-Bromo-2-chloro-1,8-naphthyridine 98%
6-Bromo-2-chloro-1,8-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₄BrClN₂
MW: 243.49 g/mol
MDL Number: MFCD07781152
CAS Number: 902837-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR