m-Tolylacetic acid 98%
Supplier: Ambeed
|
Warning
|
A153753-100G
A153753-1G
A153753-25G
A153753-5G
A153753-500G
76812-358EA
142
USD
76812-358
76812-360
76812-362
76812-364
77883-207
m-Tolylacetic acid 98%
m-Tolylacetic acid
Precaución: for Research Use Only
Formula:
(C₆H₄(CH₃))CH₂COOH |
MDL Number:
MFCD00004340 CAS Number: 621-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...