3-(Dimethylamino)-1-(naphthalen-1-yl)propan-1-one hydrochloride 97%

Supplier: Ambeed
Warning

A153557-100G A153557-10G A153557-25G A153557-5G
76812-348EA 73 USD
76812-348 76812-350 76812-352 76812-354
3-(Dimethylamino)-1-(naphthalen-1-yl)propan-1-one hydrochloride 97%
3-(Dimethylamino)-1-(naphthalen-1-yl)propan-1-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₅H₁₈ClNO
MDL Number: MFCD00554996
CAS Number: 5409-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR