5-Bromo-2-chloro-3-nitropyridine 97%

Supplier: Ambeed
Warning

A153096-100G A153096-1G A153096-500G A153096-5G A153096-25G A153096-10G
76812-318EA 32 USD
76812-318 76812-320 76812-322 76812-324 77592-872 77611-444
5-Bromo-2-chloro-3-nitropyridine 97%
5-Bromo-2-chloro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClN₂O₂
MW: 237.44
MDL Number: MFCD00222270
CAS Number: 67443-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR