N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Bis[N-phenyl-N-(m-tolyl)amino]biphenyl, N,N'-Diphenyl-N,N'-di(m-tolyl)benzidine, TPD, N,N'-Bis(3-methylphenyl)-N,N'-diphenylbenzidine

A153094-100G A153094-1G A153094-25G A153094-5G
76812-310EA 165 USD
76812-310 76812-312 76812-314 76812-316
N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine 97%
N,N'-Bis(3-methylphenyl)-N,N'-bis(phenyl)benzidine

Precaución: for Research Use Only

Formula: C₃₈H₃₂N₂
MW: 516.6741 g/mol
MDL Number: MFCD00144965
CAS Number: 65181-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR