2-Bromo-5-iodo-3-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-iodo-3-picoline

A152786-100G A152786-1G A152786-250MG A152786-25G A152786-5G
76812-296EA 736.27 USD
76812-296 76812-298 76812-300 76812-302 76812-304
2-Bromo-5-iodo-3-methylpyridine 98%
2-Bromo-5-iodo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrIN
MW: 297.92 g/mol
MDL Number: MFCD09037466
CAS Number: 65550-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR