5-Amino-1-(4-bromophenyl)-1H-pyrazole-4-carbonitrile 95%

Supplier: Ambeed
Warning

A152517-250MG A152517-5G
76812-278EA 8.49 USD
76812-278 76812-280
5-Amino-1-(4-bromophenyl)-1H-pyrazole-4-carbonitrile 95%
5-Amino-1-(4-bromophenyl)-1H-pyrazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₇BrN₄
MW: 263.09 g/mol
MDL Number: MFCD00128292
CAS Number: 5334-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR