Ethyl 6-chloro-5-methylpicolinate 97%

Supplier: Ambeed
Warning

A151949-1G A151949-250MG
76812-242EA 151.71 USD
76812-242 76812-244
Ethyl 6-chloro-5-methylpicolinate 97%
Ethyl 6-chloro-5-methylpicolinate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.63 g/mol
MDL Number: MFCD17011770
CAS Number: 178421-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR