Methyl-4-fluoro-2-methylbenzoate 98%

Supplier: Ambeed
Warning

A151933-100G A151933-10G A151933-1G A151933-25G
76812-232EA 119 USD
76812-232 76812-234 76812-236 76812-238
Methyl-4-fluoro-2-methylbenzoate 98%
Methyl 4-fluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD06203790
CAS Number: 174403-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR