(1's,4'r)-4-(3,4-Difluorophenethyl)-4'-propyl-1,1'-bi(cyclohexane) 97%

Supplier: Ambeed
Warning

A151674-1G A151674-250MG A151674-5G
76812-212EA 20.16 USD
76812-212 76812-214 76812-216
(1's,4'r)-4-(3,4-Difluorophenethyl)-4'-propyl-1,1'-bi(cyclohexane) 97%
(1's,4'r)-4-(3,4-Difluorophenethyl)-4'-propyl-1,1'-bi(cyclohexane)

Precaución: for Research Use Only

Formula: C₂₃H₃₄F₂
MW: 348.51 g/mol
MDL Number: MFCD20488042
CAS Number: 107215-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR