2-Methylpyrimidin-4-ol 98%

Supplier: Ambeed
Warning

A151324-1G A151324-250MG
76812-206EA 45.62 USD
76812-206 76812-208
2-Methylpyrimidin-4-ol 98%
2-Methylpyrimidin-4-ol

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.1139 g/mol
MDL Number: MFCD00234103
CAS Number: 67383-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR