7-Bromo-3,4-dihydroquinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A151306-250MG A151306-25G A151306-1G A151306-5G
76812-202EA 41.38 USD
76812-202 76812-204 77567-186 77671-126
7-Bromo-3,4-dihydroquinolin-2(1H)-one 97%
7-Bromo-3,4-dihydroquinolin-2(1H)-one

Precaución: For research use only.

Formula: C₉H₈BrNO
MW: 226.07 g/mol
MDL Number: MFCD09878790
CAS Number: 14548-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR