1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A151077-100MG A151077-250MG A151077-50MG
76812-190EA 89 USD
76812-190 76812-192 76812-194
1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid 95%
1-(tert-Butoxycarbonyl)-3-fluoropiperidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₈FNO₄
MDL Number: MFCD18909772
CAS Number: 1303974-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR