4-Amino-3-bromophenol 97%

Supplier: Ambeed
Warning

A150570-100MG A150570-1G A150570-250MG A150570-5G
76812-168EA 12 USD
76812-168 76812-170 76812-172 76812-174
4-Amino-3-bromophenol 97%
4-Amino-3-bromophenol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MDL Number: MFCD09966089
CAS Number: 74440-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR