PD-0173074 ≥99%

Supplier: Ambeed
Warning

A150425-10MG A150425-1MG
76812-164EA 49 USD
76812-164 76812-166
PD-0173074 ≥99%
PD-0173074

Precaución: for Research Use Only

Formula: C₂₈H₄₁N₇O₃
MDL Number: MFCD08705327
CAS Number: 219580-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR