GW-3965 hydrochloride ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
GW3965 hydrochloride, 2-(3-(3-((2-Chloro-3-(trifluoromethyl)benzyl)(2,2-diphenylethyl)amino)propoxy)phenyl)acetic acid hydrochloride, 3-[3-[[[2-Chloro-3-(trifluoromethyl)phenyl]methyl](2,2-diphenylethyl)amino]propoxy]benzeneacetic acid hydrochloride
A150197-10MG
A150197-1MG
A150197-5MG
A150197-100MG
A150197-250MG
76812-156EA
44
USD
76812-156
76812-158
76812-160
77671-114
77671-116
GW-3965 hydrochloride ≥98%
GW-3965 hydrochloride
Precaución: for Research Use Only
Formula:
C₃₃H₃₁ClF₃NO₃·HCl MW: 618.51 g/mol |
MDL Number:
MFCD08276920 CAS Number: 405911-17-3 UN: 1759 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...