Orotic acid 98+%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid

A149396-100G A149396-25G A149396-500G
76812-126EA 14.86 USD
76812-126 76812-128 76812-130
Orotic acid 98+%
Orotic acid

Precaución: for Research Use Only

Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
MDL Number: MFCD00006027
CAS Number: 65-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR