5-Amino-2-methylisonicotinic acid 97%

Supplier: Ambeed
Warning

A149357-100MG A149357-250MG A149357-1G
76812-122EA 147.47 USD
76812-122 76812-124 77611-384
5-Amino-2-methylisonicotinic acid 97%
5-Amino-2-methylisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD15143307
CAS Number: 88482-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR