Ethyl 2-(2-chloroethoxy)acetate 98%

Supplier: Ambeed
Danger

A149345-10G A149345-1G A149345-5G A149345-25G
76812-116EA 14 USD
76812-116 76812-118 76812-120 77611-382
Ethyl 2-(2-chloroethoxy)acetate 98%
Ethyl 2-(2-chloroethoxy)acetate

Precaución: for Research Use Only

Formula: C₆H₁₁ClO₃
MDL Number: MFCD08458712
CAS Number: 17229-14-0
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR