4-(Bromomethyl)benzamide 95%

Supplier: Ambeed
Danger

A149235-100MG A149235-1G A149235-250MG
76812-114EA 35 USD
76812-114 77671-100 77671-102
4-(Bromomethyl)benzamide 95%
4-(Bromomethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MDL Number: MFCD09743698
CAS Number: 58914-40-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR