4-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(3-Fluorophenylcarbamoyl)benzeneboronic acid, (4-((3-Fluorophenyl)carbamoyl)phenyl)boronic acid

A148495-1G A148495-25G A148495-5G
76812-102EA 39 USD
76812-102 76812-104 76812-106
4-(3-Fluorophenylcarbamoyl)phenylboronic acid 98%
4-(3-Fluorophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BFNO₃
MDL Number: MFCD09027204
CAS Number: 874288-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR