3-Hydroxy-2-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A148334-100MG A148334-1G A148334-250MG A148334-5G
76812-090EA 13 USD
76812-090 76812-092 76812-094 77883-175
3-Hydroxy-2-methylbenzaldehyde 98%
3-Hydroxy-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.1500 g/mol
MDL Number: MFCD09999328
CAS Number: 90111-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR