Methyl (R)-(-)-3-hydroxyisobutyrate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl (R)-(-)-beta-hydroxyisobutyrate, (R)-Methyl 3-hydroxy-2-methylpropanoate

A148151-100G A148151-10G A148151-1G A148151-5G A148151-25G
76812-078EA 499 USD
76812-078 76812-080 76812-082 76812-084 77549-820
Methyl (R)-(-)-3-hydroxyisobutyrate 97%
Methyl (R)-(-)-3-hydroxyisobutyrate

Precaución: for Research Use Only

Formula: C₅H₁₀O₃
MDL Number: MFCD00063450
CAS Number: 72657-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR