(S)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate hydrochloride 95%

Supplier: Ambeed
Warning

A147842-100MG A147842-250MG
76812-074EA 25.47 USD
76812-074 76812-076
(S)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate hydrochloride 95%
(S)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₁ClN₂O₂
MW: 236.74 g/mol
MDL Number: MFCD11101393
CAS Number: 1070968-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR