4-Cyano-3-methylbenzoic acid 95%

Supplier: Ambeed
Warning

A147447-100G A147447-10G A147447-1G A147447-250MG A147447-25G A147447-5G
76812-042EA 631 USD
76812-042 76812-044 76812-046 76812-048 76812-050 76812-052
4-Cyano-3-methylbenzoic acid 95%
4-Cyano-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MDL Number: MFCD14582961
CAS Number: 73831-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR