((Difluoromethyl)sulfonyl)benzene 98%

Supplier: Ambeed

Synonyms: Difluoromethyl phenyl sulphone

A147352-100G A147352-10G A147352-1G A147352-25G A147352-5G
76812-024EA 420 USD
76812-024 76812-026 76812-028 76812-030 76812-032
((Difluoromethyl)sulfonyl)benzene 98%
((Difluoromethyl)sulfonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₆F₂O₂S
MW: 192.18 g/mol
MDL Number: MFCD01050170
CAS Number: 1535-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR