(R)-Azepan-3-amine 95%

Supplier: Ambeed

A147278-100MG A147278-10G A147278-250MG A147278-5G A147278-1G
76812-014EA 35 USD
76812-014 76812-016 76812-018 76812-020 77671-076
(R)-Azepan-3-amine 95%
(R)-Azepan-3-amine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MDL Number: MFCD00044964
CAS Number: 124932-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR