3-Amino-2-chloroisonicotinic acid 97%

Supplier: Ambeed
Warning

A146911-10G A146911-1G A146911-250MG
76812-004EA 234 USD
76812-004 76812-006 76812-008
3-Amino-2-chloroisonicotinic acid 97%
3-Amino-2-chloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD15143308
CAS Number: 58483-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR