3,5,6-Trichloropyridin-2(1H)-one 98%

Supplier: Ambeed
Warning

A145627-100G A145627-10G A145627-1G A145627-25G A145627-5G
76811-984EA 74 USD
76811-984 76811-986 76811-988 76811-990 76811-992
3,5,6-Trichloropyridin-2(1H)-one 98%
3,5,6-Trichloropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C5H2Cl3NO
MDL Number: MFCD00130269
CAS Number: 6515-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR