(2S,3S)-(+)-2,3-Butanediol 97%

Supplier: Ambeed
Warning

A145558-100MG A145558-1G A145558-250MG A145558-25G A145558-5G A145558-100G
76811-974EA 8 USD
76811-974 76811-976 76811-978 76811-980 76811-982 77883-163
(2S,3S)-(+)-2,3-Butanediol 97%
(2S,3S)-(+)-2,3-Butanediol

Precaución: for Research Use Only

Formula: C₄H₁₀O₂
MDL Number: MFCD00063648
CAS Number: 19132-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR