2-(4,5-Dihydro-1H-imidazol-2-yl)pyridine 95%

Supplier: Ambeed
Warning

A145538-100MG A145538-250MG A145538-5G
76811-966EA 78.51 USD
76811-966 76811-968 77671-064
2-(4,5-Dihydro-1H-imidazol-2-yl)pyridine 95%
2-(4,5-Dihydro-1H-imidazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.17 g/mol
MDL Number: MFCD09743888
CAS Number: 7471-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR